ContactUsLinkComponent
Potential-free contact, 24v/2 a, with socket to namur ne 28, for signal generation, controlled by programme segment, for a total of 3 freely selectable functions to be activated, 2 contacts for unplus 1 * 1 items
Catalog # MEMMUN75PLUSH72
Supplier:  MEMMERT
Potential-free contact, 24v/2 a, with socket to namur ne 28, for signal generation, controlled by programme segment, for a total of 3 freely selectable functions to be activated, 2 contacts for unplus 1 * 1 items
Catalog # MEMMUN75PLUSH72
Supplier:  MEMMERT
Frequently Bought Together