ContactUsLinkComponent
Potential-free contact, 24 v/2 a, with socket to namur ne 28 for comination error message, e. g. supply failure, sensor fault, for i fplus 1 * 1 items
Catalog # MEMMIF450PLUSH6
Supplier:  MEMMERT
Potential-free contact, 24 v/2 a, with socket to namur ne 28 for comination error message, e. g. supply failure, sensor fault, for i fplus 1 * 1 items
Catalog # MEMMIF450PLUSH6
Supplier:  MEMMERT
Frequently Bought Together