Quartz tubing, fused, 25mm (0.984in) OD, 22mm (0.866in) ID 1 * 48 "
42274.NT
Quartz tubing, fused, 25mm (0.984in) OD, 22mm (0.866in) ID 1 * 48 "
42274.NT
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Precision tweezers, resistant, DURAX™
: OUTILS RUBIS
Carbon disulphide 99.9% for spectroscopy
: Thermo Fisher Scientific
Ethanol absolute 99.8% for analysis
: Thermo Fisher Scientific
Acetone, BAKER, J.T.Baker®
: Avantor
Toluene, extra dry 99.85%, AcroSeal®
: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
: Thermo Fisher Scientific
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals