ContactUsLinkComponent
Pestle porcelain Dia 30mm Length 135mm very good chemical resistance with rough surface at head, for mortars with ø 83 + 90 mm 1 * 2 items
Catalog # MARI6836003
Supplier:  MARIENFELD
Pestle porcelain Dia 30mm Length 135mm very good chemical resistance with rough surface at head, for mortars with ø 83 + 90 mm 1 * 2 items
Catalog # MARI6836003
Supplier:  MARIENFELD
Frequently Bought Together