ContactUsLinkComponent
Reverse osmosis membrane,spiral-membrane from composite-polyamide ,precipitating rate : 98% permeate output at 14 bar:0,38 m /day size: 2 x 13” 1 * 1 items
Catalog # WERN1932-003
Reverse osmosis membrane,spiral-membrane from composite-polyamide ,precipitating rate : 98% permeate output at 14 bar:0,38 m /day size: 2 x 13” 1 * 1 items
Catalog # WERN1932-003
Frequently Bought Together