ContactUsLinkComponent
Buffer pack PCR Phusion High Fidelity detergent-free final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer , 2x1,5ml of 50mM MgCl2 solution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-520L
Supplier:  FINNZYMES REAGENTS
Buffer pack PCR Phusion High Fidelity detergent-free final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer , 2x1,5ml of 50mM MgCl2 solution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-520L
Supplier:  FINNZYMES REAGENTS
Frequently Bought Together