ContactUsLinkComponent
22 COMPOSES A20 MG/L CHAQUE DANS HNO3 5%,HF tr Ag Al As Ba Be Ca Cd Co Cr3+ Cu Fe K Mg Mn Na Ni Pb Sb Se Tl V Zn 1 * 100 mL
Catalog # 89139.180
22 COMPOSES A20 MG/L CHAQUE DANS HNO3 5%,HF tr Ag Al As Ba Be Ca Cd Co Cr3+ Cu Fe K Mg Mn Na Ni Pb Sb Se Tl V Zn 1 * 100 mL
Catalog # 89139.180
Frequently Bought Together