ContactUsLinkComponent
ERK1/2 (THR202/TYR204) IN-CELL ELISA KIT 1 * 96 Tests
ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-Cell ELISA Kit
Catalog # ABCAAB126423-96
Supplier:  Abcam
CAS Number:  
undefined
ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-Cell ELISA Kit
Catalog # ABCAAB126423-96
Supplier:  Abcam
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-Cell ELISA kit
Specifications
About this item
ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) In-cell ELISA kit is cell-based (qualitative) ELISA for the measurement of ERK1 (phospho T202 + Y204) + ERK2 (phospho T185 + Y187) production in Mouse, Rat, Human in Adherent cells samples.
Frequently Bought Together