CONTROLLSTANDARD 10 MG/L C 1 * 1 items
Catalog # HACHLCW844
Supplier: Hach
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
STANDARD SOLUTION 50 MG/L NO3-N 1 * 1 L
Supplier: Hach
HCL ACID REAGENT 1 * 20 L
Supplier: Hach
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach