ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, S100B (MOUSE) RECOMBINANT PROTEIN, MOUSE S100B (NP_033141, 1 A.A. - 92 A.A.) FULL-LENGTH RECOMBINANT PRO TEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP4340
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, S100B (MOUSE) RECOMBINANT PROTEIN, MOUSE S100B (NP_033141, 1 A.A. - 92 A.A.) FULL-LENGTH RECOMBINANT PRO TEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP4340
Supplier:  Abnova