ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, LRPAP1 (HUMAN) RECOMBINANT PROTEIN, HUMAN LRPAP1 (NP_002328, 35 A.A. - 357 A.A.) PARTIAL RECOMBINANT PRO TEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP4907
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, LRPAP1 (HUMAN) RECOMBINANT PROTEIN, HUMAN LRPAP1 (NP_002328, 35 A.A. - 357 A.A.) PARTIAL RECOMBINANT PRO TEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP4907
Supplier:  Abnova