RECOMBINANT/NATIVE PROTEIN, H2N2 (A/JAPAN/305/1957) RECOMBINANT PROTEIN, H2N2 (A/JAPAN/305/1957, AAA43185, 17 A.A. - 529 A.A.) PARTI AL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP4897
Supplier: Abnova