ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, PEROXIDASE (HORSERADISH), EIA GRADE, LYOPHILIZED. PURIFIED. SINGLE BASIC ISOZYME WITH RZ &GT 2.9. 1 * 5.000 EU
Catalog # ABNOP5313
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, PEROXIDASE (HORSERADISH), EIA GRADE, LYOPHILIZED. PURIFIED. SINGLE BASIC ISOZYME WITH RZ &GT 2.9. 1 * 5.000 EU
Catalog # ABNOP5313
Supplier:  Abnova