ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, HA1 (H1N1) (A/SOLOMON ISLANDS/3/2006) RECOMBINANT PROTEIN, HA1 (H1N1) (A/SOLOMON ISLANDS/3/2006) (ABU990 69, 18 A.A. - 344 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP5055
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, HA1 (H1N1) (A/SOLOMON ISLANDS/3/2006) RECOMBINANT PROTEIN, HA1 (H1N1) (A/SOLOMON ISLANDS/3/2006) (ABU990 69, 18 A.A. - 344 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP5055
Supplier:  Abnova