RECOMBINANT/NATIVE PROTEIN, HA1 (H8N4) (INFLUENZA A) RECOMBINANT PROTEIN, HA1 (H8N4) (INFLUENZA A) (ABB87729, 16 A.A. - 343 A.A.) PA RTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP5050
Supplier: Abnova