ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, STREPTAVIDIN (STREPTOMYCES AVIDINII) RECOMBINANT PROTEIN, STREPTOMYCES AVIDINII STREPTAVIDIN (CAA27265, P22629, 25 A.A. - 183 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP4990
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, STREPTAVIDIN (STREPTOMYCES AVIDINII) RECOMBINANT PROTEIN, STREPTOMYCES AVIDINII STREPTAVIDIN (CAA27265, P22629, 25 A.A. - 183 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP4990
Supplier:  Abnova