ContactUsLinkComponent
KIT, METHYL-CPG BINDING DOMAIN PROTEIN 2 CHIP KIT, METHYL-CPG BINDING DOMAIN PROTEIN 2 CHIP KIT IS FOR INVESTIGATING THE INTERACTION S OF THE BINDING OF MBD2 TO METHYLATED DNA IN VIVO EFFICIENTLY. 1 * 1 KIT
Catalog # ABNOKA0694
Supplier:  Abnova
KIT, METHYL-CPG BINDING DOMAIN PROTEIN 2 CHIP KIT, METHYL-CPG BINDING DOMAIN PROTEIN 2 CHIP KIT IS FOR INVESTIGATING THE INTERACTION S OF THE BINDING OF MBD2 TO METHYLATED DNA IN VIVO EFFICIENTLY. 1 * 1 KIT
Catalog # ABNOKA0694
Supplier:  Abnova