KIT, GLOBAL PAN-METHYL HISTONE H3-K79 QUANTIFICATION KIT (FLUOROMETRIC), GLOBAL PAN-METHYL HISTONE H3-K79 QUANTIFICATION KIT (FLUORO METRIC) IS USE FOR MEASURING GLOBAL MONO, DI- AND TRI-METHYLATION OF HISTONE H3-K79. 1 * 1 KIT
Catalog # ABNOKA0623
Supplier: Abnova