KIT, GLOBAL PAN-METHYL HISTONE H3-K36 QUANTIFICATION KIT (FLUOROMETRIC), GLOBAL PAN-METHYL HISTONE H3-K36 QUANTIFICATION KIT (FLUORO METRIC) IS USE FOR MEASURING GLOBAL MONO-, DI- AND TRIMETHYLATION OF HISTONE H3-K36. 1 * 1 KIT
Catalog # ABNOKA0615
Supplier: Abnova