ContactUsLinkComponent
KIT, GLOBAL PAN-METHYL HISTONE H3-K9 QUANTIFICATION KIT (COLORIMETRIC), GLOBAL PAN-METHYL HISTONE H3-K9 QUANTIFICATION KIT (COLORIME TRIC) IS USE FOR MEASURING GLOBAL MONO-, DI- AND TRI-METHYLATION OF HISTONE H3-K9. 1 * 1 KIT
Catalog # ABNOKA0598
Supplier:  Abnova
KIT, GLOBAL PAN-METHYL HISTONE H3-K9 QUANTIFICATION KIT (COLORIMETRIC), GLOBAL PAN-METHYL HISTONE H3-K9 QUANTIFICATION KIT (COLORIME TRIC) IS USE FOR MEASURING GLOBAL MONO-, DI- AND TRI-METHYLATION OF HISTONE H3-K9. 1 * 1 KIT
Catalog # ABNOKA0598
Supplier:  Abnova