ContactUsLinkComponent
KIT, CXCL9 (HUMAN) ELISA KIT, CXCL9 (HUMAN) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF HUMAN CXC L9. 1 * 1 KIT
Catalog # ABNOKA0555
Supplier:  Abnova
KIT, CXCL9 (HUMAN) ELISA KIT, CXCL9 (HUMAN) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF HUMAN CXC L9. 1 * 1 KIT
Catalog # ABNOKA0555
Supplier:  Abnova
Frequently Bought Together