ContactUsLinkComponent
KIT, SS-B AB ELISA KIT, SS-B AB ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QUANTITATIVE MEASUREMENT OF IGG CLASS AUTOANTIBOD IES AGAINST SS-B (LA) IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1116
Supplier:  Abnova
KIT, SS-B AB ELISA KIT, SS-B AB ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QUANTITATIVE MEASUREMENT OF IGG CLASS AUTOANTIBOD IES AGAINST SS-B (LA) IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1116
Supplier:  Abnova