ContactUsLinkComponent
KIT, SS-A AB ELISA KIT, SS-A AB ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QUANTITATIVE MEASUREMENT OF IGG CLASS AUTOANTIBOD IES AGAINST SS-A (RO, 52 AND 60 KDA) IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
  ABNOKA1115
 :  Abnova
KIT, SS-A AB ELISA KIT, SS-A AB ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR THE QUANTITATIVE MEASUREMENT OF IGG CLASS AUTOANTIBOD IES AGAINST SS-A (RO, 52 AND 60 KDA) IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
  ABNOKA1115
 :  Abnova
Frequently Bought Together