0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, 25 MM MGCL2, 25 MM MGCL2 CAN BE USED FOR OPTIMIZATION OF MAGNESIUM ION CONCENTRATION IN PCR REAC TION. 1 * 1,8 mL
Catalog # ABNOD0023
Supplier:
Abnova
Expand image
PRODUCT FOR DNA RELATE APPLICATION, 25 MM MGCL2, 25 MM MGCL2 CAN BE USED FOR OPTIMIZATION OF MAGNESIUM ION CONCENTRATION IN PCR REAC TION. 1 * 1,8 mL
Catalog # ABNOD0023
Supplier:
Abnova
Frequently Bought Together
cDNA synthesis kits, AffinityScript
Supplier: AGILENT
SCS110 competent cells
Supplier: AGILENT
RNA isolation, total RNA kit, E-Z 96®
Supplier: OMEGA BIO-TEK