ContactUsLinkComponent
ANTIBODY ARRAY, ERK SIGNALING PHOSPHO-SPECIFIC ARRAY, ERK SIGNALING PHOSPHO-SPECIFIC ARRAY INCLUDES 208 HIGHLY SPECIFIC AND WELL-CHA RACTERIZED PHOSPHORYLATION ANTIBODIES IN THE ERK PATHWAY. 1 * 2 items
  ABNOAA0025
 :  Abnova
ANTIBODY ARRAY, ERK SIGNALING PHOSPHO-SPECIFIC ARRAY, ERK SIGNALING PHOSPHO-SPECIFIC ARRAY INCLUDES 208 HIGHLY SPECIFIC AND WELL-CHA RACTERIZED PHOSPHORYLATION ANTIBODIES IN THE ERK PATHWAY. 1 * 2 items
  ABNOAA0025
 :  Abnova