ContactUsLinkComponent
ANTIBODY ARRAY, AKT/PKB SIGNALING PHOSPHO-SPECIFIC ARRAY, AKT/PKB SIGNALING PHOSPHO-SPECIFIC ARRAY INCLUDES 216 HIGHLY SPECIFIC AND WELL-CHARACTERIZED PHOSPHORYLATION ANTIBODIES IN THE AKT/PKB SIGNALING PATHWAY. 1 * 2 items
Catalog # ABNOAA0013
Supplier:  Abnova
ANTIBODY ARRAY, AKT/PKB SIGNALING PHOSPHO-SPECIFIC ARRAY, AKT/PKB SIGNALING PHOSPHO-SPECIFIC ARRAY INCLUDES 216 HIGHLY SPECIFIC AND WELL-CHARACTERIZED PHOSPHORYLATION ANTIBODIES IN THE AKT/PKB SIGNALING PATHWAY. 1 * 2 items
Catalog # ABNOAA0013
Supplier:  Abnova