ContactUsLinkComponent
SOLUTION DE SODIUM CHROMATE 0,005M Sodium Chromate 0.005M/ 2.5% Myristyltrimethylammonium bromide/ 0.04% Boric acid 1 * 500 mL
Catalog # CMPLFC435670
SOLUTION DE SODIUM CHROMATE 0,005M Sodium Chromate 0.005M/ 2.5% Myristyltrimethylammonium bromide/ 0.04% Boric acid 1 * 500 mL
Catalog # CMPLFC435670
Frequently Bought Together