NUTRIENT AGAR ISO 16266 1 * 100 g
Catalog # LIOF620036
Supplier: LIOFILCHEM
Frequently Bought Together
VWR®, Ladies' Coats, Cotton or Polyester/Cotton
Supplier: VWR Collection
Dilution Rack for Compact Dry
Supplier: HYSERVE
Dehydrated culture media
Supplier: Merck
VWR® Culture Media, Ready to Use, in Bottles
Supplier: VWR Chemicals
PLATE COUNT AGAR 1 * 100 g
Supplier: LIOFILCHEM
Media in Bottles
Supplier: MERCK LIFE SCIENCE
Orange Serum Agar
Supplier: LIOFILCHEM
VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
YEAST NITROGEN BASE, FOR MICROBIOLOGY* 1 * 100 g
Supplier: Merck
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
Ready Culture Media in Tubes
Supplier: LIOFILCHEM