ContactUsLinkComponent
Optical nitrate probe with spectral processing of UVVIS range, for in-situ measurement in the effluent of municipal wastewater treatment plants, with integrated ultrasonic cleaning. Supplied with multifunctional slide and Shock-Absorption-Rings, wit 1 * 1 items
Catalog # WTWO481046
Supplier:  WTW ONLINE
Optical nitrate probe with spectral processing of UVVIS range, for in-situ measurement in the effluent of municipal wastewater treatment plants, with integrated ultrasonic cleaning. Supplied with multifunctional slide and Shock-Absorption-Rings, wit 1 * 1 items
Catalog # WTWO481046
Supplier:  WTW ONLINE
Frequently Bought Together