ContactUsLinkComponent
Recombinant protein, lrrc57( np_694992.1, 1 a.a. - 239 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00255252-P01
Supplier:  Abnova
Recombinant protein, lrrc57( np_694992.1, 1 a.a. - 239 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00255252-P01
Supplier:  Abnova