ContactUsLinkComponent
Recombinant protein, h1foo( aah47943.1, 1 a.a. - 207 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00132243-P01
Supplier:  Abnova
Recombinant protein, h1foo( aah47943.1, 1 a.a. - 207 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00132243-P01
Supplier:  Abnova