ContactUsLinkComponent
Recombinant protein, mon1a( np_115731.1, 1 a.a. - 555 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00084315-P01
Supplier:  Abnova
Recombinant protein, mon1a( np_115731.1, 1 a.a. - 555 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00084315-P01
Supplier:  Abnova