ContactUsLinkComponent
Recombinant protein, flj14166( aah13353.1, 1 a.a. - 121 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00079616-P01
Supplier:  Abnova
Recombinant protein, flj14166( aah13353.1, 1 a.a. - 121 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00079616-P01
Supplier:  Abnova