ContactUsLinkComponent
Recombinant protein, c16orf70( np_079463.2, 1 a.a. - 422 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00080262-P01
Supplier:  Abnova
Recombinant protein, c16orf70( np_079463.2, 1 a.a. - 422 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00080262-P01
Supplier:  Abnova