ContactUsLinkComponent
Recombinant protein, suds3( np_071936.2, 1 a.a. - 328 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00064426-P01
Supplier:  Abnova
Recombinant protein, suds3( np_071936.2, 1 a.a. - 328 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00064426-P01
Supplier:  Abnova