ContactUsLinkComponent
Recombinant protein, klf3( np_057615, 1 a.a. - 69 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00051274-Q01
Supplier:  Abnova
Recombinant protein, klf3( np_057615, 1 a.a. - 69 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00051274-Q01
Supplier:  Abnova