ContactUsLinkComponent
Recombinant protein, sez6l( cak54604.1, 1 a.a. - 948 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00023544-P01
Supplier:  Abnova
undefined
Recombinant protein, sez6l( cak54604.1, 1 a.a. - 948 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00023544-P01
Supplier:  Abnova