ContactUsLinkComponent
Recombinant protein, csrp3( np_003467, 95 a.a. - 194 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008048-Q01
Supplier:  Abnova
Recombinant protein, csrp3( np_003467, 95 a.a. - 194 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008048-Q01
Supplier:  Abnova