ContactUsLinkComponent
Recombinant protein, pry( np_004667.2, 48 a.a. - 147 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009081-Q01
Supplier:  Abnova
undefined
Recombinant protein, pry( np_004667.2, 48 a.a. - 147 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009081-Q01
Supplier:  Abnova