ContactUsLinkComponent
Recombinant protein, mad2l1bp( np_055443.1, 171 a.a. - 274 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009587-Q01
Supplier:  Abnova
undefined
Recombinant protein, mad2l1bp( np_055443.1, 171 a.a. - 274 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009587-Q01
Supplier:  Abnova