ContactUsLinkComponent
Recombinant protein, nte( np_006693.2, 1229 a.a. - 1327 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010908-Q01
Supplier:  Abnova
undefined
Recombinant protein, nte( np_006693.2, 1229 a.a. - 1327 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010908-Q01
Supplier:  Abnova