ContactUsLinkComponent
Recombinant protein, trim16( np_006461, 165 a.a. - 273 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010626-Q01
Supplier:  Abnova
Recombinant protein, trim16( np_006461, 165 a.a. - 273 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010626-Q01
Supplier:  Abnova