ContactUsLinkComponent
Recombinant protein, ccl18( np_002979, 21 a.a. - 89 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006362-Q01
Supplier:  Abnova
undefined
Recombinant protein, ccl18( np_002979, 21 a.a. - 89 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006362-Q01
Supplier:  Abnova