ContactUsLinkComponent
Recombinant protein, ccl14( aah45165, 20 a.a. - 93 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006358-Q01
Supplier:  Abnova
Recombinant protein, ccl14( aah45165, 20 a.a. - 93 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006358-Q01
Supplier:  Abnova