ContactUsLinkComponent
Recombinant protein, apoc3( np_000031.1, 21 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000345-Q01
Supplier:  Abnova
Recombinant protein, apoc3( np_000031.1, 21 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000345-Q01
Supplier:  Abnova