ContactUsLinkComponent
Recombinant protein, slc25a6( np_001627, 135 a.a. - 209 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000293-Q01
Supplier:  Abnova
Recombinant protein, slc25a6( np_001627, 135 a.a. - 209 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000293-Q01
Supplier:  Abnova