ContactUsLinkComponent
Recombinant protein, adar( np_001102.2, 2 a.a. - 110 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00000103-Q01
Supplier:  Abnova
Recombinant protein, adar( np_001102.2, 2 a.a. - 110 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00000103-Q01
Supplier:  Abnova