ContactUsLinkComponent
Recombinant protein, acaca( np_942131, 1 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000031-Q01
Supplier:  Abnova
undefined
Recombinant protein, acaca( np_942131, 1 a.a. - 99 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000031-Q01
Supplier:  Abnova