ContactUsLinkComponent
Recombinant protein, f9( np_000124, 96 a.a. - 190 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002158-Q01
Supplier:  Abnova
undefined
Recombinant protein, f9( np_000124, 96 a.a. - 190 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002158-Q01
Supplier:  Abnova