ContactUsLinkComponent
Recombinant protein, ids( np_000193, 38 a.a. - 137 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003423-Q01
Supplier:  Abnova
Recombinant protein, ids( np_000193, 38 a.a. - 137 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003423-Q01
Supplier:  Abnova